Thermo Scientific Chemicals

Trifluoroacetic acid, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 044630.AP
500 mL, Each
Thermo Scientific Chemicals

Trifluoroacetic acid, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 044630.AP
500 mL, Each
Quantity
Catalog number: 044630.AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-05-1
IUPAC Name
trifluoroacetic acid
Molecular Formula
C2HF3O2
InChI Key
DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)F
Form
Liquid
Optical Absorbance at
210 nm, 1.00 AU, max
Appearance (Color)
Clear colorless
Assay (Aqueous acid-base Titration)
≥99.5%
Optical Absorbance at
230 nm, 0.15 AU, max

Description

The combination of properties like solubility in most of the solvents, volatility, catalytic property and strong acidity with non-oxidizing nature makes it a widely used reagent in an organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and ceramic compounds. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. It finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text