Trifluoroacetic acid, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Trifluoroacetic acid, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 500 mL
500 mL
Catalog number 044630.AY
also known as 044630-AY
Price (USD)/ Each
915.65
Online exclusive
1017.00 
Save 101.35 (10%)
-
Add to cart
Quantity:
4 x 500 mL
Request bulk or custom format
Price (USD)/ Each
915.65
Online exclusive
1017.00 
Save 101.35 (10%)
Add to cart
Trifluoroacetic acid, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Catalog number044630.AY
Price (USD)/ Each
915.65
Online exclusive
1017.00 
Save 101.35 (10%)
-
Add to cart
Chemical Identifiers
CAS76-05-1
IUPAC Nametrifluoroacetic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Optical Absorbance at210 nm, 1.00 AU, max
Appearance (Color)Clear colorless
Assay (Aqueous acid-base Titration)≥99.5%
Optical Absorbance at230 nm, 0.15 AU, max
View more
The combination of properties like solubility in most of the solvents, volatility, catalytic property and strong acidity with non-oxidizing nature makes it a widely used reagent in an organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and ceramic compounds. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. It finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only