Thermo Scientific Chemicals

Cesium trifluoroacetate, Thermo Scientific Chemicals

Catalog number: 044633.18
50 g, Each
Thermo Scientific Chemicals

Cesium trifluoroacetate, Thermo Scientific Chemicals

Catalog number: 044633.18
50 g, Each
Quantity
Catalog number: 044633.18
also known as 044633-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21907-50-6
IUPAC Name
caesium(1+) trifluoroacetate
Molecular Formula
C2CsF3O2
InChI Key
RJYSYRSELCQCSO-UHFFFAOYSA-M
SMILES
[Cs+].[O-]C(=O)C(F)(F)F
Assay (unspecified)
98% min

Description

It is used in acid catalyzed reactions, especially those where an ester is cleaved in peptide synthesis. Used as an ion pairing agent in liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text