Thermo Scientific Chemicals
ICP-MS Internal Standard solution, Specpure™, Thermo Scientific Chemicals
Catalog number: 044651.AE
100 mL, Each
Thermo Scientific Chemicals
ICP-MS Internal Standard solution, Specpure™, Thermo Scientific Chemicals
Catalog number: 044651.AE
100 mL, Each
Quantity
Catalog number: 044651.AE
also known as 044651-AE
Price (USD)
Have Questions?
Concentration
₆Li 100?g/mL ?2%
Concentration
Bi 100?g/mL ?2%
Concentration
In 100?g/mL ?2%
Concentration
Tb 100?g/mL ?2%
Concentration
Sc 100?g/mL ?2%