Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals
Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 044661.22
also known as 044661-22
Price (USD)/ Each
216.65
Online exclusive
241.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
216.65
Online exclusive
241.00 
Save 24.35 (10%)
Add to cart
Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals
Catalog number044661.22
Price (USD)/ Each
216.65
Online exclusive
241.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1308-06-1
IUPAC Namedicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular FormulaCo3O4
InChI KeyUBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only