Thermo Scientific Chemicals

Poly(acrylic acid), 25 wt% soln. in water, Thermo Scientific Chemicals

Catalog number: 044669.22
100 g, Each
Thermo Scientific Chemicals

Poly(acrylic acid), 25 wt% soln. in water, Thermo Scientific Chemicals

Catalog number: 044669.22
100 g, Each
Quantity
Catalog number: 044669.22
also known as 044669-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-01-4
IUPAC Name
Poly(acrylic acid)
Molecular Formula
(C3H4O2)n
SMILES
OC(=O)C(-*)C-*
MDL Number
MFCD00084394
Form
Liquid
Assay from Suppliers CofA
23.0-27.0% w/w [Poly(acrylic acid)]
Clarity
Clear
Identification (FTIR)
Conforms

Description

Polyacrylic acid is used in disposable diapers and in ion exchange resins. It is also used to study solute diffusion in polyvinyl alcohol/polyacrylic acid copolymer hydrogel. It is also employed as a thickening, suspending, emulsifying and dispersing agent in pharmaceuticals, cosmetics, adhesives and paints. Further, it is used for the preparation of poly(N-isopropylacrylamide)-block-polyacrylic acid copolymer which responds to both temperature and pH stimuli. In addition to this, it is used in preparing block copolymer of oligo (methyl methacrylate)/PAA for micellar delivery of hydrophobic drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text