Ethyl hydrogen malonate, 90+%, Thermo Scientific Chemicals
Ethyl hydrogen malonate, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl hydrogen malonate, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Catalog number 044692.36
also known as 044692-36
Price (USD)/ Each
1,496.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,496.00
Add to cart
Ethyl hydrogen malonate, 90+%, Thermo Scientific Chemicals
Catalog number044692.36
Price (USD)/ Each
1,496.00
-
Add to cart
Specifications
Chemical Name or MaterialEthyl hydrogen malonate
CAS1071-46-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Ethyl hydrogen malonate is used as a reactant for the preparation of tetramic acids through Dieckmann ring closure and organocatalytic decarboxylative Doebner-Knoevenagel reactions. It is involved in the acylation reactions and Knoevenagel condensation with aldehydes. It is also used in the preparation of gamma-lactones from olefins by intermolecular carbolactonization in presence of Mn(III) acetate as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only