Thermo Scientific Chemicals

Soda lime, indicating, ACS, Thermo Scientific Chemicals

Catalog number: 044697.36
500 g, Each
Thermo Scientific Chemicals

Soda lime, indicating, ACS, Thermo Scientific Chemicals

Catalog number: 044697.36
500 g, Each
Quantity
Catalog number: 044697.36
also known as 044697-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8006-28-8
IUPAC Name
sodium oxocalcium hydroxide
Molecular Formula
CaHNaO2
InChI Key
HUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES
[OH-].[Na+].O=[Ca]
Comment
1% Fines
Loss on Drying
7%
Comment
19.0% min CO₂ absorption capacity

Description

Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text