Soda lime, indicating, ACS, Thermo Scientific Chemicals
Soda lime, indicating, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Soda lime, indicating, ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Catalog number 044697.A4
also known as 044697-A4
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
Add to cart
Soda lime, indicating, ACS, Thermo Scientific Chemicals
Catalog number044697.A4
Price (USD)/ Each
284.65
Online exclusive
316.00 
Save 31.35 (10%)
-
Add to cart
Chemical Identifiers
CAS8006-28-8
IUPAC Namesodium oxocalcium hydroxide
Molecular FormulaCaHNaO2
InChI KeyHUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES[OH-].[Na+].O=[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment1% Fines
Loss on Drying7%
Comment19.0% min CO₂ absorption capacity
Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only