Thermo Scientific Chemicals

Poly(styrene-divinylbenzene), 2% cross-linked, 200-400 mesh, Thermo Scientific Chemicals

Catalog number: 044708.36
500 g, Each
Thermo Scientific Chemicals

Poly(styrene-divinylbenzene), 2% cross-linked, 200-400 mesh, Thermo Scientific Chemicals

Catalog number: 044708.36
500 g, Each
Quantity
Catalog number: 044708.36
also known as 044708-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-70-7
MDL Number
9003-70-7
Form
Powder/Beads
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Particle Size
200-400 Mesh: Conforms
Volume
Swollen Volume: 5.0-7.0 mL/g in DCM

Description

Styrene-divinylbenzene Copolymer is used as packing material (solid phase) to determine the contamination of groundwater using solid phase extraction based method. It is widely used as column packing material in liquid chromatography for separation of both organic and inorganic ions.3 Styrene-divinylbenzene Copolymer is used in inorganic trace analysis for solid phase extraction preconcentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text