Thermo Scientific Chemicals

Magnesium oxide, nanopowder, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 044733.36
500 g, Each
Thermo Scientific Chemicals

Magnesium oxide, nanopowder, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 044733.36
500 g, Each
Quantity
Catalog number: 044733.36
also known as 044733-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-48-4
IUPAC Name
oxomagnesium
Molecular Formula
MgO
InChI Key
CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES
O=[Mg]
Form
Powder
Total Metal Impurities
≤1%
Surface Area
≥30m²/g
Appearance (Color)
White
Particle Size
40-60nm

Description

Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, as a protective coating in plasma displays, as an oxide barrier in spin-tunneling devices, as an optical material, as a component of tubular construction in heating elements and also in libraries for preserving books. Magnesium oxide microspheres are used in the enrichment method of polycyclic aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text