Thermo Scientific Chemicals

Hydrogen tetrachloroaurate(III), solution, Au 40-44% w/w (cont. Au), Thermo Scientific Chemicals

Catalog number: 044744.NH
1 g, Each
Thermo Scientific Chemicals

Hydrogen tetrachloroaurate(III), solution, Au 40-44% w/w (cont. Au), Thermo Scientific Chemicals

Catalog number: 044744.NH
1 g, Each
Quantity
Catalog number: 044744.NH
also known as 044744-NH
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16903-35-8
IUPAC Name
gold(3+) ammonium tetrachloride
Molecular Formula
AuCl4H4N
InChI Key
WPEJSSRSFRWYJB-UHFFFAOYSA-K
SMILES
[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Au+3]
Assay (unspecified)
40-44% Au

Description

Hydrogen tetrachloroaurate(III) acts as a precursor used in the purification of gold by electrolysis. It is also used for the recovery, concentration, purification and analytical determination of gold. It acts as a key starting material for the preparation of gold compounds. It plays an important role as a catalyst in the preparation of 2-arylbenzoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text