Thermo Scientific Chemicals

Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals

Catalog number: 044745.02
0.5 g, Each
Thermo Scientific Chemicals

Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals

Catalog number: 044745.02
0.5 g, Each
Quantity
Catalog number: 044745.02
Price (USD)
Have Questions?

Chemical Identifiers

CAS
104934-50-1
Molecular Formula
(C10H14S)n
SMILES
CCCCCCC1=C(-*)SC(-*)=C1
MDL Number
MFCD00217686
Molecular Weight
20,000 - 70,000
Trace Metals (by AAS)
Ni < 100 ppm

Description

Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) and devices. It is also used as organic light emitting diode (OLED) materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text