Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals
Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044745.77
Price (USD)/ Each
-
Quantity:
0.1 g
Request bulk or custom format
Chemical Identifiers
CAS104934-50-1
Molecular Formula(C10H14S)n
SMILESCCCCCCC1=C(-*)SC(-*)=C1
MDL NumberMFCD00217686
SpecificationsSpecification SheetSpecification Sheet
Molecular Weight20,000 - 70,000
Trace Metals (by AAS)Ni < 100 ppm
Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) and devices. It is also used as organic light emitting diode (OLED) materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only