Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, nanopowder, 99+%, Thermo Scientific Chemicals

Catalog number: 044757.14
25 g, Each
Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, nanopowder, 99+%, Thermo Scientific Chemicals

Catalog number: 044757.14
25 g, Each
Quantity
Catalog number: 044757.14
also known as 044757-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Form
Powder
Total Metal Impurities
≤ 1.0%
Surface Area
100-200m²/g
Appearance (Color)
White
Particle Size
20nm Average

Description

Aluminum oxide is used in protein extraction as a component in the preparation of tissues. It is also used as a grinding or blending agent. It can act as a catalyst in a variety of reactions such as the Claus process. It is used in the production of aluminum, manufacture of abrasives, refractories, ceramics, electrical insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text