Aluminum oxide, gamma-phase, nanopowder, 99+%, Thermo Scientific Chemicals
Aluminum oxide, gamma-phase, nanopowder, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, nanopowder, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044757.36
also known as 044757-36
Price (USD)/ Each
551.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
649.00 
Save 97.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
551.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
649.00 
Save 97.35 (15%)
Add to cart
Aluminum oxide, gamma-phase, nanopowder, 99+%, Thermo Scientific Chemicals
Catalog number044757.36
Price (USD)/ Each
551.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
649.00 
Save 97.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤ 1.0%
Surface Area100-200m²/g
Appearance (Color)White
Particle Size20nm Average
Aluminum oxide is used in protein extraction as a component in the preparation of tissues. It is also used as a grinding or blending agent. It can act as a catalyst in a variety of reactions such as the Claus process. It is used in the production of aluminum, manufacture of abrasives, refractories, ceramics, electrical insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only