2,2'-(Ethylenedioxy)bis(ethylamine), 97+%, Thermo Scientific Chemicals
2,2'-(Ethylenedioxy)bis(ethylamine), 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2'-(Ethylenedioxy)bis(ethylamine), 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 044759.09
also known as 044759-09
Price (USD)/ Each
24.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
24.50
Add to cart
2,2'-(Ethylenedioxy)bis(ethylamine), 97+%, Thermo Scientific Chemicals
Catalog number044759.09
Price (USD)/ Each
24.50
-
Add to cart
Chemical Identifiers
CAS929-59-9
IUPAC Name2-[2-(2-aminoethoxy)ethoxy]ethan-1-amine
Molecular FormulaC6H16N2O2
InChI KeyIWBOPFCKHIJFMS-UHFFFAOYSA-N
SMILESNCCOCCOCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 97.0%
FormLiquid
Appearance (Color)Clear, colorless
Refractive Index1.4590 - 1.4630
2,2'-(Ethylenedioxy)bis(ethylamine) is used to linker to form conjugates of manganese-doped zinc sulfide nanoparticles with folic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only