Thermo Scientific Chemicals

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 044770.30
250 g, Each
Thermo Scientific Chemicals

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 044770.30
250 g, Each
Quantity
Catalog number: 044770.30
also known as 044770-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23519-77-9
IUPAC Name
tetrapropoxyzirconium
Molecular Formula
C12H28O4Zr
InChI Key
XPGAWFIWCWKDDL-UHFFFAOYSA-N
SMILES
CCCO[Zr](OCCC)(OCCC)OCCC
Form
Liquid
Assay from Suppliers CofA
ZrO2 Content: 27.0-29.0%
Specific Gravity
1.050-1.090 g/mL

Description

Zirconium(IV) n-propoxide is used as a precursor for the preparation of Zirconia. It is used as coupling agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text