(+)-Biotin N-succinimidyl ester, 98%, Thermo Scientific Chemicals
(+)-Biotin N-succinimidyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+)-Biotin N-succinimidyl ester, 98%, Thermo Scientific Chemicals

In coupling reactions to amino acids, peptides and proteins through primary amines
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 044771.01
also known as 044771-01
Price (USD)/ Each
183.65
Online exclusive
204.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
0.25 g
Request bulk or custom format
Price (USD)/ Each
183.65
Online exclusive
204.00 
Save 20.35 (10%)
Add to cart
(+)-Biotin N-succinimidyl ester, 98%, Thermo Scientific Chemicals
Catalog number044771.01
Price (USD)/ Each
183.65
Online exclusive
204.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS35013-72-0
IUPAC Name2,5-dioxopyrrolidin-1-yl 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoate
Molecular FormulaC14H19N3O5S
InChI KeyYMXHPSHLTSZXKH-UHFFFAOYNA-N
SMILESO=C(CCCCC1SCC2NC(=O)NC12)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥97.5%
Appearance (Color)White to cream
Optical Rotation+51.0 to +54.0°
(+)-Biotin N-succinimidyl ester is used in coupling reactions to amino acids, peptides and proteins through primary amines. It is used as amino reactive biotin reagent and in the preparation of biotinylated surfaces or polypeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only