Thermo Scientific Chemicals

(+)-Biotin N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 044771.03
1 g, Each
Thermo Scientific Chemicals

(+)-Biotin N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 044771.03
1 g, Each
Quantity
Catalog number: 044771.03
also known as 044771-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35013-72-0
IUPAC Name
2,5-dioxopyrrolidin-1-yl 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoate
Molecular Formula
C14H19N3O5S
InChI Key
YMXHPSHLTSZXKH-UHFFFAOYNA-N
SMILES
O=C(CCCCC1SCC2NC(=O)NC12)ON1C(=O)CCC1=O
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥97.5%
Appearance (Color)
White to cream
Optical Rotation
+51.0 to +54.0°

Description

(+)-Biotin N-succinimidyl ester is used in coupling reactions to amino acids, peptides and proteins through primary amines. It is used as amino reactive biotin reagent and in the preparation of biotinylated surfaces or polypeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text