Thermo Scientific Chemicals
ICP-MS Internal Standard solution for 200.8 & CLP ILM05.2,Specpure™, Thermo Scientific Chemicals
Catalog number: 044789.AE
100 mL, Each
Thermo Scientific Chemicals
ICP-MS Internal Standard solution for 200.8 & CLP ILM05.2,Specpure™, Thermo Scientific Chemicals
Catalog number: 044789.AE
100 mL, Each
Quantity
Catalog number: 044789.AE
also known as 044789-AE
Price (USD)
Have Questions?
Concentration
Bi 10?g/mL ?2%
Concentration
Lu 10?g/mL ?2%
Concentration
Tb 10?g/mL ?2%
Concentration
Y 10?g/mL ?2%
Concentration
Sc 10?g/mL ?2%