Ammonium tungsten oxide hydrate, Thermo Scientific Chemicals
Ammonium tungsten oxide hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium tungsten oxide hydrate, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 044792.36
also known as 044792-36
Price (USD)/ Each
281.65
Online exclusive
313.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
281.65
Online exclusive
313.00 
Save 31.35 (10%)
Add to cart
Ammonium tungsten oxide hydrate, Thermo Scientific Chemicals
Catalog number044792.36
Price (USD)/ Each
281.65
Online exclusive
313.00 
Save 31.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12333-11-8
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Ammonium tungsten oxide hydrate is widely utilized as a raw material for the synthesis of tungsten catalysts, which is used in a variety of reactions like oxidation, hydroxylation, hydrogenation and polymerization. It is used as a reagent for chemical analysis like medical diagnosis. It is used as a precursor material for the preparation of high-purity WO3 which is used as thin-film substrate for certain semiconductor devices. It acts as a precursor to a variety of POM (polyoxometalates).