Thermo Scientific Chemicals

Ethyl carbamate, 98%, Thermo Scientific Chemicals

Catalog number: 044804.30
250 g, Each
Thermo Scientific Chemicals

Ethyl carbamate, 98%, Thermo Scientific Chemicals

Catalog number: 044804.30
250 g, Each
Quantity
Catalog number: 044804.30
also known as 044804-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
51-79-6
IUPAC Name
ethyl carbamate
Molecular Formula
C3H7NO2
InChI Key
JOYRKODLDBILNP-UHFFFAOYSA-N
SMILES
CCOC(N)=O
Assay (GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤0.5%
Form
Crystalline powder
Assay from Supplier's CofA
Chloride: ≤0.01%
Sulfated Ash
≤0.1%

Description

Ethyl carbamate in its molten state is good solvent for various organic materials, preparation & modification of amino resins, as solubilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text