Thermo Scientific Chemicals
Platinum Iridium wire, 1.5mm (0.06in) dia, Thermo Scientific Chemicals
Catalog number: 044812.BU
25 cm, Each
Thermo Scientific Chemicals
Platinum Iridium wire, 1.5mm (0.06in) dia, Thermo Scientific Chemicals
Catalog number: 044812.BU
25 cm, Each
Quantity
Catalog number: 044812.BU
also known as 044812-BU
Price (USD)
Quantity
-
Have Questions?
Description
Platinum-iridium wire is used in platinum jewelry, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibers; in metrology and in fabrication of metal microelectrodes. It is used in the electrical stimulation of nervous tissue and in electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.