Thermo Scientific Chemicals

Cyclopentane, HPLC Grade, Thermo Scientific Chemicals

Catalog number: 044814.K2
1 L, Each
Thermo Scientific Chemicals

Cyclopentane, HPLC Grade, Thermo Scientific Chemicals

Catalog number: 044814.K2
1 L, Each
Quantity
Catalog number: 044814.K2
also known as 044814-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
287-92-3
IUPAC Name
cyclopentane
Molecular Formula
C5H10
InChI Key
RGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILES
C1CCCC1
Assay (GC)
>/= 99.8% as sum of C5 + C6 hydrocarbons
Residual water
0.02%
Optical Absorbance at
240 nm 0.01 AU, max
Optical Absorbance at
210 nm 0.50 AU, max
Residue on Evaporation
3ppm

Description

Cyclopentane is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text