5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%
5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%
5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%
5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%
Thermo Scientific Chemicals

5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%

CAS: 40615-39-2 | C31H32N2O7 | 544.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 044815.02
also known as 044815-02
Price (USD)
135.65
Online Exclusive
150.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
135.65
Online Exclusive
150.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS40615-39-2
IUPAC Name1-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC31H32N2O7
InChI KeyUBTJZUKVKGZHAD-QZGLRKMJNA-N
SMILESCOC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C)C(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder or crystals
Assay from Suppliers CofA≥98.0% (HPLC)
Water Content (Karl Fischer Titration)≤1.5%
Residual solvents≤5.0%
Ask our AI about this Product
5'-O-(4,4'-Dimethoxytrityl)thymidine is used in the solid-phase synthesis of polynucleotides and polythymidylic acids by the block coupling phosphotriester method. Further, it is used in the stereoselective synthesis of 3'-deoxy-3'-threo-hydroxymethyl nucleoside. In addition to this, it is used as a research tool for antiviral and anticancer studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link