Thermo Scientific Chemicals
5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%, Thermo Scientific Chemicals
Catalog number: 044815.04
2 g, Each
Thermo Scientific Chemicals
5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%, Thermo Scientific Chemicals
Catalog number: 044815.04
2 g, Each
Quantity
Catalog number: 044815.04
also known as 044815-04
Price (USD)
Quantity
-
Have Questions?
Description
5'-O-(4,4'-Dimethoxytrityl)thymidine is used in the solid-phase synthesis of polynucleotides and polythymidylic acids by the block coupling phosphotriester method. Further, it is used in the stereoselective synthesis of 3'-deoxy-3'-threo-hydroxymethyl nucleoside. In addition to this, it is used as a research tool for antiviral and anticancer studies.