Niobium(V) oxalate hydrate, Thermo Scientific Chemicals
Niobium(V) oxalate hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium(V) oxalate hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 044819.22
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
Add to cart
Niobium(V) oxalate hydrate, Thermo Scientific Chemicals
Catalog number044819.22
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1660137-09-6
IUPAC Nametetrakis[(carboxycarbonyl)oxy]niobio hydrogen oxalate
Molecular FormulaC10H5NbO20
InChI KeyAXIFGFAGYFPNFC-UHFFFAOYSA-I
SMILESOC(=O)C(=O)O[Nb](OC(=O)C(O)=O)(OC(=O)C(O)=O)(OC(=O)C(O)=O)OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)19.9% Nb₂O₅ min
Niobium(V) oxalate hydrate is used as doping precursors in the PZT composition to improve its piezoelectric properties. It is also used in the preparation of niobium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only