Nickel foil, 0.1mm (0.004in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
Nickel foil, 0.1mm (0.004in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel foil, 0.1mm (0.004in) thick, 99.5% (metals basis), Thermo Scientific Chemicals

Form: Approx.2.2g/50 x 50mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 500 mm
50 x 50 mm
100 x 100 mm
Catalog number 044821.HP
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
100 x 500 mm
Request bulk or custom format
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
Add to cart
Nickel foil, 0.1mm (0.004in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number044821.HP
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver-grey
FormFoil (0.1 mm thickness)
Total Metal Impurities≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only