Nickel foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Nickel foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Form: Approx.5.6g/50 x 50mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 044822.FI
also known as 044822-FI
Price (USD)/ Each
30.60
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
30.60
Add to cart
Nickel foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number044822.FI
Price (USD)/ Each
30.60
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormFoil, 0.25mm (0.01in) thick, annealed.
Assay from Supplier's CofA≥99.5% (metals basis)
Assay from Supplier's CofA99.5% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only