Nickel foil, 0.5mm (0.02in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Nickel foil, 0.5mm (0.02in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel foil, 0.5mm (0.02in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Form: Approx.11.1g/50 x 50mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 044823.FI
also known as 044823-FI
Price (USD)/ Each
34.90
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
34.90
Add to cart
Nickel foil, 0.5mm (0.02in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number044823.FI
Price (USD)/ Each
34.90
-
Add to cart
Specifications
Chemical Name or MaterialNickel
ColorSilver to Gray
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 3P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only