Thermo Scientific Chemicals

Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%, Thermo Scientific Chemicals

Catalog number: 044829.03
1 g, Each
Thermo Scientific Chemicals

Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%, Thermo Scientific Chemicals

Catalog number: 044829.03
1 g, Each
Quantity
Catalog number: 044829.03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95408-45-0
IUPAC Name
Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
Molecular Formula
C26H44Cl2FeP2Pd
SMILES
[Fe].Cl[Pd]Cl.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C
Molecular Weight (g/mol)
651.75
Elemental Analysis
Wt % Cl 10.13 to 11.63
Elemental Analysis
Wt% Carbon 46.91 to 48.91
Elemental Analysis
Wt% H 6.05 to 7.55

Description

Coupling reactions, Suzuki couplings.Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene] palladium(II) is used as a catalyst in Suzuki-cross coupling reactions, which involve reaction of aromatic halides with methyliminodiacetic acid derivatives. it is also used as a catalyst for the preparation of chalcones. It is used in the preparation of (bisphosphinometallocene)-palladium(II) chloride complexes, which used as a starting material for the Buchwald-Hartwig reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text