Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%, Thermo Scientific Chemicals
Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%, Thermo Scientific Chemicals

Coupling reactions, Suzuki couplings
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.25 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044829.77
Price (USD)/ Each
-
Quantity:
0.1 g
Request bulk or custom format
Chemical Identifiers
CAS95408-45-0
IUPAC NameDichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
Molecular FormulaC26H44Cl2FeP2Pd
SMILES[Fe].Cl[Pd]Cl.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C
Molecular Weight (g/mol)651.75
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisWt % Cl 10.13 to 11.63
Elemental AnalysisWt% Carbon 46.91 to 48.91
Elemental AnalysisWt% H 6.05 to 7.55
Coupling reactions, Suzuki couplings.Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene] palladium(II) is used as a catalyst in Suzuki-cross coupling reactions, which involve reaction of aromatic halides with methyliminodiacetic acid derivatives. it is also used as a catalyst for the preparation of chalcones. It is used in the preparation of (bisphosphinometallocene)-palladium(II) chloride complexes, which used as a starting material for the Buchwald-Hartwig reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only