Thermo Scientific Chemicals

Indium(III) sulfide, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 044835.04
2 g, Each
Thermo Scientific Chemicals

Indium(III) sulfide, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 044835.04
2 g, Each
Quantity
Catalog number: 044835.04
also known as 044835-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12030-24-9
IUPAC Name
diindium(3+) trisulfanediide
Molecular Formula
In2S3
InChI Key
SIXIBASSFIFHDK-UHFFFAOYSA-N
SMILES
[S--].[S--].[S--].[In+3].[In+3]
Total Metal Impurities
0.02% max.

Description

Indium(III) sulfide acts as mid band gap semiconductor. It is an important material for optoelectronic and photovoltaic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text