Thermo Scientific Chemicals

Bis(tri-tert-butylphosphine)palladium(0), Thermo Scientific Chemicals

Catalog number: 044845.01
0.25 g, Each
Thermo Scientific Chemicals

Bis(tri-tert-butylphosphine)palladium(0), Thermo Scientific Chemicals

Catalog number: 044845.01
0.25 g, Each
Quantity
Catalog number: 044845.01
also known as 044845-01
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53199-31-8
IUPAC Name
bis(tri-tert-butylphosphane) palladium
Molecular Formula
C24H54P2Pd
InChI Key
MXQOYLRVSVOCQT-UHFFFAOYSA-N
SMILES
[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
Elemental Analysis
Wt % H 10.10 to 11.15
Elemental Analysis
Wt % C 55.40 to 57.40

Description

Bis(tri-tert-butylphosphine)palladium(0) is used in Coupling reactions, Heck couplings. It acts as a palladium catalyst for reductive carbonylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under inert atmosphere
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text