2-Methyl-1-pyrroline, Thermo Scientific Chemicals
2-Methyl-1-pyrroline, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-1-pyrroline, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 mL
Catalog number 044853.AC
also known as 044853-AC
Price (USD)/ Each
278.00
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
278.00
Add to cart
2-Methyl-1-pyrroline, Thermo Scientific Chemicals
Catalog number044853.AC
Price (USD)/ Each
278.00
-
Add to cart
Chemical Identifiers
CAS872-32-2
IUPAC Name2-methylpyrrolidin-2-ylium
Molecular FormulaC5H10N
InChI KeyOZLJZWIVBYLSOK-UHFFFAOYSA-N
SMILESC[C+]1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Colorless to yellow
FormLiquid
2-Methyl-1-pyrroline acts as monocyclic imine. It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only