Thermo Scientific Chemicals

Iodine lump, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044857.22
100 g, Each
Thermo Scientific Chemicals

Iodine lump, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044857.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 044857.22
also known as 044857-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7553-56-2
IUPAC Name
diiodine
Molecular Formula
I2
InChI Key
PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES
II
Total Metal Impurities
0.001% max.

Description

Iodine lump is used in OLED materials, pharmaceuticals, and the electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text