Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max, Thermo Scientific Chemicals
Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 044879.06
also known as 044879-06
Price (USD)/ Each
541.65
Online exclusive
602.00 
Save 60.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
541.65
Online exclusive
602.00 
Save 60.35 (10%)
Add to cart
Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max, Thermo Scientific Chemicals
Catalog number044879.06
Price (USD)/ Each
541.65
Online exclusive
602.00 
Save 60.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Ca ≤ 100ppm
Total Metal Impurities≤0.01%
Particle Size-200 mesh
FormPowder
Assay (special)Ru 99.8-100.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only