Thermo Scientific Chemicals

Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals

Catalog number: 044895.14
25 g, Each
Thermo Scientific Chemicals

Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals

Catalog number: 044895.14
25 g, Each
Quantity
Catalog number: 044895.14
also known as 044895-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1309-37-1
IUPAC Name
trioxodiiron
Molecular Formula
Fe2O3
InChI Key
JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES
[Fe][Fe](=O)(=O)=O
Form
Powder
Surface Area
30-60 m²/g
Particle Size
20-40 nm (conforms)
Appearance (Color)
Red

Description

NanoArc cosmetic iron oxide is an auburn material offering unique cosmetic qualities. The small particle size results in a strong color development combined with high translucency for a unique glowing look.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 30-60m2/g
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text