Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals
Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals

Nanoarc Cosmetic Iron Oxide Is An Auburn Material offering Unique Cosmetic Qualitites. The Small Particle Size Results in a Strong Color Development Combined With High Translucency, For a Unique Glowing Look
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044895.36
also known as 044895-36
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1309-37-1
IUPAC Nametrioxodiiron
Molecular FormulaFe2O3
InChI KeyJEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES[Fe][Fe](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red or brown
FormPowder
Particle Size20-40 nm (conforms)
Surface Area30-60 m²/g
NanoArc cosmetic iron oxide is an auburn material offering unique cosmetic qualities. The small particle size results in a strong color development combined with high translucency for a unique glowing look.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 30-60m2/g
RUO – Research Use Only