Iron(III) oxide, magnetic, NanoArc™, Thermo Scientific Chemicals
Iron(III) oxide, magnetic, NanoArc™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(III) oxide, magnetic, NanoArc™, Thermo Scientific Chemicals

Nanoarc Magnetic Iron Oxide Is a Highly Magnetic Material in Which the Crystallites Are >98% Gamma-pHase. This Material Is Suitable For a Variety of Applications Requiring Strong Magnetic Properties and Narrow Particle
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044896.14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1309-37-1
IUPAC Nametrioxodiiron
Molecular FormulaFe2O3
InChI KeyJEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES[Fe][Fe](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to red-brown to brown
FormPowder
Particle Size20-40nm
Surface Area30-60m²/g
X-Ray Diffraction (XRD)Gamma: Conforms
Iron(III) oxide acts as a pigment, in paints, coatings and colored concretes, used in thermalite. Iron oxide is a low cost, nontoxic, ecofriendly catalyst for various reactions such as the oxidation of carbon monoxide, reduction of nitrogen oxides and sulfur dioxide by CO, photocatalytic splitting of water, Fischer-Tropsch synthesis of hydrocarbons, catalytic decomposition of industrial dyes, water-gas shift reactions, nucleation of diamonds, conversion of phenolic and other environmentally harmful aromatic compounds, dehydrogenation of ethylbenzene to styrene and catalytic oxidations of various organic compounds. It is used in magnetic storage devices and in the production of iron, steel and alloys. It functions as a catalyst in the chemical oxidation of hydrogen peroxide used for the treatment of contaminated water with hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 30-60m2/g
RUO – Research Use Only