Thermo Scientific Chemicals

Lead(II) selenide, 99.999+% (metals basis), Thermo Scientific Chemicals

Catalog number: 044916.22
100 g, Each
Thermo Scientific Chemicals

Lead(II) selenide, 99.999+% (metals basis), Thermo Scientific Chemicals

Catalog number: 044916.22
100 g, Each
Quantity
Catalog number: 044916.22
also known as 044916-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12069-00-0
IUPAC Name
plumbaneselone
Molecular Formula
PbSe
InChI Key
GGYFMLJDMAMTAB-UHFFFAOYSA-N
SMILES
[Se]=[Pb]
Total Metal Impurities
0.001% max.

Description

Lead(II) selenide is used in the production of infrared detectors for thermal imaging. It is also used as quantum dots in nanocrystal solar cell. It is an important material and finds applications in photographic plates, and photo resistors and optoelectronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text