Zinc oxide, NanoArc™ ZN-0625, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant, Thermo Scientific Chemicals
Zinc oxide, NanoArc™ ZN-0625, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc oxide, NanoArc™ ZN-0625, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant, Thermo Scientific Chemicals

Colloidal dispersion with dispersant
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2 kg
Catalog number 044924.22
also known as 044924-22
Price (USD)/ Each
35.60
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
35.60
Add to cart
Zinc oxide, NanoArc™ ZN-0625, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant, Thermo Scientific Chemicals
Catalog number044924.22
Price (USD)/ Each
35.60
-
Add to cart
Specifications
Chemical Name or MaterialZinc oxide, NanoTek™ Z1102PMA
Name Note50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant
CAS1314-13-2
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
View more
Zinc oxide dispersion in Dowanol PMA suitable for incorporation in a wide variety of coatings formulations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zinc oxide dispersion in Dowanol™ PMA suitable for incorporation in a wide variety of coatings formulations
RUO – Research Use Only