Thermo Scientific Chemicals

Indium tin oxide, NanoTek|r, 99.5%, Thermo Scientific Chemicals

Catalog number: 044927.14
25 g, Each
Thermo Scientific Chemicals

Indium tin oxide, NanoTek|r, 99.5%, Thermo Scientific Chemicals

Catalog number: 044927.14
25 g, Each
Quantity
Catalog number: 044927.14
also known as 044927-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50926-11-9
IUPAC Name
stannanone indium
Molecular Formula
InOSn
InChI Key
AMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES
[In].O=[Sn]
Assay (unspecified)
99.5% % In₂O₃/SnO₂ min
Surface Area
10-35m²/g

Description

Indium tin oxide is one of the most widely used transparent conducting oxides due to its important properties like electrical conductivity and optical transparency, as well as the easy formation of thin film on deposition. It is also used in touch panels, electrochromic, electroluminescent and LCD displays, plasma displays, field emission displays, heat reflective coatings, energy efficient windows, gas sensors and photovoltaics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • NanoTek™ indium tin oxide can be used to impart conductivity to transparent coatings such as those on the surface of computer display tubes or on the films used to produce conductive display screens such as for interactive touch screens
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text