Zinc rod, 8mm (0.31in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals
Zinc rod, 8mm (0.31in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc rod, 8mm (0.31in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mm
50 mm
Catalog number 044936.DT
also known as 044936-DT
Price (USD)/ Each
1,985.00
-
Add to cart
Quantity:
250 mm
Request bulk or custom format
Price (USD)/ Each
1,985.00
Add to cart
Zinc rod, 8mm (0.31in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number044936.DT
Price (USD)/ Each
1,985.00
-
Add to cart
Specifications
Chemical Name or MaterialZinc rod
Name Note8mm (0.31 in.) dia.
CAS7440-66-6
Recommended StorageAmbient temperatures
EINECS Number231-175-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only