Nitinol foil, 0.127mm (0.005in) thick, superelastic, flat annealed, pickled surface, Thermo Scientific Chemicals
Nitinol foil, 0.127mm (0.005in) thick, superelastic, flat annealed, pickled surface, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitinol foil, 0.127mm (0.005in) thick, superelastic, flat annealed, pickled surface, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 044952.FI
also known as 044952-FI
Price (USD)/ Each
276.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
276.00
Add to cart
Nitinol foil, 0.127mm (0.005in) thick, superelastic, flat annealed, pickled surface, Thermo Scientific Chemicals
Catalog number044952.FI
Price (USD)/ Each
276.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialNitinol foil
Name Note0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Innovations in the manufacturing and processing of Nitinol have led to a great expansion of Nitinol medical devices in a variety of therapeutic areas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only