Lead foil, 0.76mm (0.03in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Lead foil, 0.76mm (0.03in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead foil, 0.76mm (0.03in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Form: Approx.208g/15 x 15cm
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 30 cm
15 x 15 cm
Catalog number 044961.RF
also known as 044961-RF
Price (USD)/ Each
72.65
Online exclusive
80.60 
Save 7.95 (10%)
-
Add to cart
Quantity:
30 x 30 cm
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.60 
Save 7.95 (10%)
Add to cart
Lead foil, 0.76mm (0.03in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number044961.RF
Price (USD)/ Each
72.65
Online exclusive
80.60 
Save 7.95 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialLead
ColorGray
CAS7439-92-1
Health Hazard 1H350-H360FD-H362-H372
Health Hazard 3P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only