Tantalum disilicide, Thermo Scientific Chemicals
Tantalum disilicide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum disilicide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044965.09
also known as 044965-09
Price (USD)/ Each
73.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
73.10
Add to cart
Tantalum disilicide, Thermo Scientific Chemicals
Catalog number044965.09
Price (USD)/ Each
73.10
-
Add to cart
Chemical Identifiers
CAS12039-79-1
IUPAC Name(silanylidenetantalio)silane
Molecular FormulaH4Si2Ta
InChI KeyJSCUKKCIAVANLH-UHFFFAOYSA-N
SMILES[SiH2]=[Ta]=[SiH2]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max
Tantalum disilicide is used in the preparation of ceramics, which finds application in the thermal protection systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only