Thermo Scientific Chemicals

Chlorobis(cyclooctene)iridium(I) dimer, Ir nominally 42.9%, Thermo Scientific Chemicals

Catalog number: 044966.06
5 g, Each
Thermo Scientific Chemicals

Chlorobis(cyclooctene)iridium(I) dimer, Ir nominally 42.9%, Thermo Scientific Chemicals

Catalog number: 044966.06
5 g, Each
Quantity
Catalog number: 044966.06
also known as 044966-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12246-51-4
IUPAC Name
tetrakis(cyclooctene) diiridium dichloride
Molecular Formula
C32H56Cl2Ir2
InChI Key
CJJIQMGSHWWMCK-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ir].[Ir].C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1
Assay (unspecified)
5.80-6.80% Hydrogen
Assay (unspecified)
6.90-8.90% Chlorine
Assay (unspecified)
41.90-43.90% Carbon

Description

Chlorobis(cyclooctene)iridium(I) dime is used as catalyst for Isomerization-hydroboration reactions with nido-carboranyldiphosphine as stabilizing ligand, Hydrogen peroxide oxidation of hydroxamic acids and their subsequent hetero Diels-Alder cycloaddition reactions, Immobilization of organic functional groups onto solid supports through vinylsilane coupling reactions, Alkylation reactions, Guerbet reaction, Allylic amination reactions in a DNA-diene-iridium(I) hybrid system, Asymmetric hydroamination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text