Thermo Scientific Chemicals

Methoxy(cyclooctadiene)iridium(I) dimer, Ir nominally 58%, Thermo Scientific Chemicals

Catalog number: 044967.03
1 g, Each
Thermo Scientific Chemicals

Methoxy(cyclooctadiene)iridium(I) dimer, Ir nominally 58%, Thermo Scientific Chemicals

Catalog number: 044967.03
1 g, Each
Quantity
Catalog number: 044967.03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12148-71-9
IUPAC Name
bis((1Z,5Z)-cycloocta-1,5-diene); bis(methoxyiridiumylium)
Molecular Formula
C18H30Ir2O2
InChI Key
SGBSCFWMCJNCRM-MIXQCLKLSA-N
SMILES
CO[Ir+].CO[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Assay (unspecified)
Ir 57.00-58.50%

Description

Methoxy(cyclooctadiene)iridium(I) dimer is used as a catalyst in the preparation of heteroaryl fused indole ring systems and borylation and Suzuki-miyaura coupling. Further, it is used in tetraborylation reactions, ortho-silylation of aryl ketone, benzaldehyde and benzyl alcohol derivatives through C-H activation. The most useful application is highly regio and enantio selection asymmetric hydroboration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text